Indium arsenide ≥99.9999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

12932-04 12932-09
CAAA12932-04EA 237 CAD
CAAA12932-04 CAAA12932-09 CAAA12932-18
Indium arsenide ≥99.9999% (metals basis)
Indium arsenide

MDL: MFCD00016144 Notes: Hardly attacked by mineral acids


Purity: 99.9999% CAS No.: 1303-11-3 Molecular Formula: InAs Formula Weight: 189.74 Physical Form: 1.5-9.5mm Polycrystalline Pieces Odor: Odorless Merck Reference: 14,4949 UN No.: 1557 MDL No.: MFCD00016144

Formula: AsIn
MW: 189.74 g/mol
Melting Pt: 936 °C
Density: 5.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016144
CAS Number: 1303-11-3
EINECS: 215-115-3
UN: 1557
ADR: 6.1,III
Merck Index: 14,4949

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR