Molybdenum(VI) oxide ≥99.9995% (metals basis excluding W), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Molybdenum trioxide

12930-14 12930-22 12930-36
CAAA12930-14EA 195.92 CAD
CAAA12930-14 CAAA12930-22 CAAA12930-36
Molybdenum(VI) oxide ≥99.9995% (metals basis excluding W), Puratronic®
Molybdenum (VI) oxide

MDL: MFCD00003469


Purity: 99.9995% CAS No.: 1313-27-5 Molecular Formula: MoO3 Formula Weight: 143.94 Physical Form: Powder Density: 4.692 Appearance: White to grey Odor: Odorless Melting Point: 795° Boiling Point: 1155° subl. Merck Reference: 14,6239 UN No.: 3288 MDL No.: MFCD00003469

Formula: MoO₃
MW: 143.94 g/mol
Boiling Pt: 1155 °C (subl.)
Melting Pt: 795 °C
Density: 4.692
Storage Temperature: Ambient
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1 III
Merck Index: 14,6239

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR