Cesium acetate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Acetic acid cesium salt
12929-14
12929-22
CAAA12929-14EA
121.34
CAD
CAAA12929-14
CAAA12929-22
Cesium acetate ≥99.9% (metals basis)
Cesium acetate
MDL: MFCD00013056
Purity: 99.9%
CAS No.: 3396-11-0
Molecular Formula: CsOOCCH3
Formula Weight: 191.95
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00013056
Formula:
H₃CCOOCs MW: 191.95 g/mol Melting Pt: 194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013056 CAS Number: 3396-11-0 EINECS: 222-248-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...