Cesium bromide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cesiumbromide
12928-18
12928-30
CAAA12928-18EA
167.48
CAD
CAAA12928-18
CAAA12928-30
Cesium bromide ≥99.9% (metals basis)
Cesium bromide
MDL: MFCD00010954
Purity: 99.9%
CAS No.: 7787-69-1
Molecular Formula: BrCs
Formula Weight: 212.81
Physical Form: Crystalline
Density: 4.44
Refractive Index: 1.6984
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 636°
Boiling Point: 1300°
Merck Reference: 14,2009
MDL No.: MFCD00010954
Formula:
BrCs MW: 212.81 Boiling Pt: 1300 Melting Pt: 636 Density: 4.44 Storage Temperature: Ambient |
MDL Number:
MFCD00010954 CAS Number: 7787-69-1 EINECS: 232-130-0 Merck Index: 14,2009 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...