Dysprosium trinitrate pentahydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dysprosium (III) nitrate pentahydrate
12922-14
12922-22
CAAA12922-14EA
98.12
CAD
CAAA12922-14
CAAA12922-22
Dysprosium trinitrate pentahydrate ≥99.9% (REO, rare earth oxide basis)
Dysprosium trinitrate pentahydrate
MDL: MFCD00149683
Purity: 99.9%
CAS No.: 10031-49-9
Molecular Formula: Dy(NO3)3.5H2O
Formula Weight: 438.59 (348.51anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Cream to pale yellow
Odor: Odorless
pH: 3
Melting Point: 88.6°
Merck Reference: 14,3482
UN No.: 1477
MDL No.: MFCD00149683
Formula:
DyN₃O₉·5H₂O MW: 438.59 Melting Pt: 88.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149683 CAS Number: 10031-49-9 EINECS: 233-410-5 UN: 1477 ADR: 5.1 III Merck Index: 14,3482 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...