Erbium trinitrate pentahydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12920-14
12920-22
CAAA12920-14EA
137.3
CAD
CAAA12920-14
CAAA12920-22
Erbium trinitrate pentahydrate ≥99.9% (REO, rare earth oxide basis)
Erbium trinitrate pentahydrate
MDL: MFCD00010988
Purity: 99.9%
Molecular Formula: Er(NO3)3.xH2O (x≈5)
Formula Weight: 353.27(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point: 130° -4H2O
UN No.: 1477
MDL No.: MFCD00010988
Formula:
ErN₃O₉·5H₂O Melting Pt: 130 °C (-4H₂O) Storage Temperature: Ambient |
MDL Number:
MFCD00149691 CAS Number: 10031-51-3 EINECS: 233-436-7 UN: 1477 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...