Gadolinium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
GADOLINIUM NITRATE, Gadolinium(III) nitrate hydrate
12917-14
12917-22
CAAA12917-14EA
91.01
CAD
CAAA12917-14
CAAA12917-22
Gadolinium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis)
Gadolinium(III) nitrate hydrate
MDL: MFCD00011023
Purity: 99.9%
CAS No.: 94219-55-3
Molecular Formula: Gd(NO3)3.xH2O (x≈6)
Formula Weight: 343.26(anhy)
Physical Form: Crystalline
Density: 2.406
Appearance: White
Melting Point: 92°
Merck Reference: 14,4326
UN No.: 1477
MDL No.: MFCD00011023
Formula:
Gd(NO₃)₃·xH₂O MW: 343.26 g/mol Melting Pt: 92 °C Density: 2.406 Storage Temperature: Ambient |
MDL Number:
MFCD00149730 CAS Number: 94219-55-3 EINECS: 233-437-2 UN: 1477 ADR: 5.1 III Merck Index: 14,4326 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...