Lanthanum(III) chloride heptahydrate ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
12916-18
12916-30
CAAA12916-18EA
115.5
CAD
CAAA12916-18
CAAA12916-30
Lanthanum(III) chloride heptahydrate ACS
Lanthanum (III) chloride heptahydrate
MDL: MFCD00149756
CAS No.: 10025-84-0
Molecular Formula: LaCl3.7H2O
Formula Weight: 371.37 (245.27anhy)
Physical Form: Crystalline
Odor: Odorless
Melting Point: 91° dec.
MDL No.: MFCD00149756
Formula:
LaCl₃·7H₂O Boiling Pt: ∼860 °C (1013 hPa) Melting Pt: 91 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149756 CAS Number: 10025-84-0 EINECS: 233-237-5 Merck Index: 14,5363 |
Specification Test Results
Insoluble matter | 0.01% |
Ca | 10 ppm |
MG | 10 ppm |
LaCl3 | 64.5-70.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...