Neodymium trinitrate hexahydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12912-18
12912-30
CAAA12912-18EA
126.24
CAD
CAAA12912-18
CAAA12912-30
Neodymium trinitrate hexahydrate ≥99.9% (REO, rare earth oxide basis)
Neodymium trinitrate hexahydrate
MDL: MFCD00149799
Purity: 99.9%
CAS No.: 16454-60-7
Molecular Formula: Nd(NO3)3.6H2O
Formula Weight: 438.34 (330.25anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
pH: 3.0-4.0
Melting Point: ca 40°
Merck Reference: 14,6450
UN No.: 1477
MDL No.: MFCD00149799
Formula:
N₃NdO₉·6H₂O Melting Pt: ∼40 °C Density: 2.265 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149799 CAS Number: 16454-60-7 EINECS: 233-153-9 UN: 1477 ADR: 5.1,III Merck Index: 14,6450 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...