Samarium trinitrate hexahydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Samarium (III) nitrate hexahydrate, Samarium(III) nitrate hydrate

12906-14 12906-22
CAAA12906-14EA 134.3 CAD
CAAA12906-14 CAAA12906-22
Samarium trinitrate hexahydrate ≥99.9% (REO, rare earth oxide basis)
Samarium trinitrate hexahydrate

MDL: MFCD00149857


Purity: 99.9% CAS No.: 13759-83-6 Molecular Formula: Sm(NO3)3.6H2O Formula Weight: 444.45 (336.36anhy) Physical Form: Crystalline Density: 2.375 Appearance: Pale yellow Odor: Odorless pH: ca. 3 Melting Point: 78-79° UN No.: 1477 MDL No.: MFCD00149857

Formula: N₃O₉Sm·6H₂O
MW: 444.47 g/mol
Melting Pt: 78…79 °C
Density: 2,375
Storage Temperature: Ambient
MDL Number: MFCD00149857
CAS Number: 13759-83-6
EINECS: 233-798-6
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR