Yttrium (III) nitrate hexahydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

12898-06 12898-14 12898-22 12898-59
CAAA12898-06EA 62.88 CAD
CAAA12898-06 CAAA12898-14 CAAA12898-22 CAAA12898-59
Yttrium (III) nitrate hexahydrate ≥99.9% (REO, rare earth oxide basis)
Yttrium (III) nitrate hexahydrate

MDL: MFCD00149940


Purity: 99.9% CAS No.: 13494-98-9 Molecular Formula: Y(NO3)3.6H2O Formula Weight: 383.01 (274.92anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,10107 UN No.: 1477 MDL No.: MFCD00149940

Formula: N₃O₉Y·6H₂O
Melting Pt: 51.8 °C
Density: 2.682 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149940
CAS Number: 13494-98-9
EINECS: 233-802-6
UN: 1477
ADR: 5.1,III
Merck Index: 14,10107

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR