Gadolinium ≥99% (REO, rare earth oxide basis), powder -40 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
12896-04
12896-09
12896-18
CAAA12896-04EA
78.05
CAD
CAAA12896-04
CAAA12896-09
CAAA12896-18
Gadolinium ≥99% (REO, rare earth oxide basis), powder -40 mesh
Gadolinium
MDL: MFCD00011022
Purity: 99%
CAS No.: 7440-54-2
Formula Weight: 157.25
Physical Form: Packaged under argon
Appearance: Silver
UN No.: 3178
MDL No.: MFCD00011022
Formula:
Gd Boiling Pt: 3273 °C (1013 hPa) Melting Pt: 1312 °C Density: 7.886 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011022 CAS Number: 7440-54-2 EINECS: 231-162-2 UN: 3178 ADR: 4.1,II Merck Index: 12,04346 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...