Samarium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
12895-03
12895-09
12895-18
CAAA12895-03EA
107.12
CAD
CAAA12895-03
CAAA12895-09
CAAA12895-18
Samarium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Samarium
MDL: MFCD00011233
Purity: 99.9%
CAS No.: 7440-19-9
Physical Form: Ampouled under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Silver
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00011233
Formula:
Sm Storage Temperature: Ambient |
MDL Number:
MFCD00011233 CAS Number: 7440-19-9 EINECS: 231-128-7 UN: 3089 ADR: 4.1,II Merck Index: 12,08498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...