Rubidium bromide ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
12891-22
12891-06
12891-14
CAAAA12891-22EA
715.74
CAD
CAAAA12891-22
CAAA12891-06
CAAA12891-14
Rubidium bromide ≥99.8% (metals basis)
Rubidium bromide
MDL: MFCD00011186
Soluble in water
Purity: 99.8%
CAS No.: 7789-39-1
Molecular Formula: RbBr
Formula Weight: 165.38
Physical Form: Crystalline
Refractive Index: 1.5530
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 693°
Boiling Point: 1340°
Solubility: Soluble in water
Merck Reference: 14,8283
MDL No.: MFCD00011186
Formula:
BrRb Boiling Pt: 1340 °C Melting Pt: 693 °C Density: 3.35 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011186 CAS Number: 7789-39-1 EINECS: 232-162-5 Merck Index: 14,8283 |
Specification Test Results
Form | Crystalline |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...