Rubidium acetate ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
12890-09
12890-18
CAAA12890-09EA
108.55
CAD
CAAA12890-09
CAAA12890-18
Rubidium acetate ≥99.8% (metals basis)
Rubidium acetate
MDL: MFCD00013058
Purity: 99.8%
CAS No.: 563-67-7
Molecular Formula: RbOOCCH3
Formula Weight: 144.52
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00013058
Formula:
H₃CCOORb Melting Pt: 246 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013058 CAS Number: 563-67-7 EINECS: 209-255-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...