Cesium carbonate ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

12887-06 12887-18 12887-30 12887-A1
CAAA12887-06EA 37.13 CAD
CAAA12887-06 CAAA12887-18 CAAA12887-30 CAAA12887-A1
Cesium carbonate ≥99% (metals basis)
Cesium carbonate

Catalyst in ethylene oxide polymerization, in coating spatter-free welding of steel in CO2, and in oxide cathodes


Purity: 99% CAS No.: 534-17-8 Molecular Formula: Cs2CO3 Formula Weight: 325.82 Physical Form: Powder/Granules Density: 4.072 Appearance: White Odor: Odorless pH: 10-12 Melting Point: 610° dec. Solubility: Very soluble in water and alcohol. Soluble in ether Beilstein Reference: 4546405 Merck Reference: 14,2010 MDL No.: MFCD00010957


Catalyst in ethylene oxide polymerization, in coating spatter-free welding of steel in CO2, and in oxide cathodes

Formula: Cs₂CO₃
Melting Pt: 610 °C (dec.)
Density: 4.072
Storage Temperature: Ambient
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9
Merck Index: 14,2010

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR