Ammonium hexachloroplatinate(IV) Pt ≥ 43,4%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diammonium hexachloroplatinate

12882-03 12882-06
CAAA12882-03EA 248.06 CAD
CAAA12882-03 CAAA12882-06
Ammonium hexachloroplatinate(IV) Pt ≥ 43,4%
Ammonium hexachloroplatinate(IV)

Electroplating of platinum, manufacturing of platinum sponge


CAS No.: 16919-58-7 Molecular Formula: (NH4)2PtCl6 Formula Weight: 443.89 Physical Form: Powder Density: 3.06 Sensitivity: Hygroscopic Appearance: Yellow Melting Point: dec. Solubility: Slightly soluble in water. Insoluble in alcohol Merck Reference: 14,548 UN No.: 3288 MDL No.: MFCD00010886


Electroplating of platinum, manufacturing of platinum sponge

Formula: H₈Cl₆N₂Pt
MW: 443.87
Melting Pt: Dec.
Density: 3.06
Storage Temperature: Ambient
MDL Number: MFCD00010886
CAS Number: 16919-58-7
EINECS: 240-973-0
UN: 3288
ADR: 6.1 III
Merck Index: 14,548

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR