Strontium sulfate ≥99.9965% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
12870-06
12870-14
CAAA12870-06EA
122.13
CAD
CAAA12870-06
CAAA12870-14
Strontium sulfate ≥99.9965% (metals basis), Puratronic®
Strontium sulphate
MDL: MFCD00036274
Purity: 99.9965%
CAS No.: 7759-02-6
Molecular Formula: O4SSr
Formula Weight: 183.68
Physical Form: Powder
Density: 3.960
Appearance: White
Odor: Odorless
Melting Point: 1600°
Merck Reference: 14,8848
MDL No.: MFCD00036274
Formula:
O₄SSr Melting Pt: 1600 °C Density: 3.960 Storage Temperature: Ambient |
MDL Number:
MFCD00036274 CAS Number: 7759-02-6 EINECS: 231-850-2 Merck Index: 14,8848 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...