Platinum ≥99.95% (metals basis), thin foil, Thickness: 0.004+/-0.0025 mm (0.00016+/-0.0001 in)

Supplier: Thermo Scientific Chemicals

12868-FF 12868-FI
CAAA12868-FFEA 605.14 CAD
CAAA12868-FF CAAA12868-FI
Platinum ≥99.95% (metals basis), thin foil, Thickness: 0.004+/-0.0025 mm (0.00016+/-0.0001 in)
Platinum

Physical Form: ∼0.05 g/25x25 mm, not certified pinhole free


Purity: 99.95% CAS No.: 7440-06-4 Physical Form: ≈0.05g/25x25mm, not certified pinhole free Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR