Zinc oxide ≥99.9995% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
12855-14
12855-22
CAAA12855-14EA
254.38
CAD
CAAA12855-14
CAAA12855-22
Zinc oxide ≥99.9995% (metals basis), Puratronic®
Zinc oxide
MDL: MFCD00011300
Purity: 99.9995%
CAS No.: 1314-13-2
Molecular Formula: ZnO
Formula Weight: 81.37
Physical Form: Powder
Density: 5.606
Melting Point: 1975°
Merck Reference: 14,10147
UN No.: 3077
MDL No.: MFCD00011300
Formula:
ZnO Melting Pt: 1975 °C Density: 5.606 Storage Temperature: Ambient |
MDL Number:
MFCD00011300 CAS Number: 1314-13-2 EINECS: 215-222-5 UN: 3077 ADR: 9,III REACH: 01-2119463881-32 Merck Index: 14,10147 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...