Aluminium sulfide 99+% (metals basis), pieces
Supplier: Thermo Scientific Chemicals
|
Warning
|
12851-06
12851-14
CAAA12851-06EA
314.42
CAD
CAAA12851-06
CAAA12851-14
Aluminium sulfide 99+% (metals basis), pieces
Aluminium sulphide
MDL: MFCD00014162
Purity: 99+%
CAS No.: 1302-81-4
Molecular Formula: Al2S3
Formula Weight: 150.16
Physical Form: 0.64cm (0.25in) & down pieces
Density: 2.02
Appearance: Pale gray
Odor: Like rotten eggs
Melting Point: 1100°
Merck Reference: 14,367
UN No.: 3134
MDL No.: MFCD00014162
Formula:
Al₂S₃ Boiling Pt: 1500 °C (1013 hPa) Melting Pt: 1100 °C Density: 2.02 Storage Temperature: Ambient |
MDL Number:
MFCD00014162 CAS Number: 1302-81-4 EINECS: 215-109-0 UN: 3134 ADR: 4.3,III Merck Index: 14,367 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...