Diarsenic triselenide ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Arsenic(III) selenide
12849-06
12849-14
CAAA12849-02EA
115.92
CAD
CAAA12849-02
CAAA12849-06
CAAA12849-14
Diarsenic triselenide ≥99.999% (metals basis)
Diarsenic triselenide
For vacuum deposition
Purity: 99.999%
CAS No.: 1303-36-2
Molecular Formula: As2Se3
Formula Weight: 386.72
Physical Form: 1-6mm Fused Pieces
Density: 4.75
Melting Point: 260°
Merck Reference: 14,805
UN No.: 3077
MDL No.: MFCD00014166
For vacuum deposition
Formula:
As₂Se₃ Melting Pt: 260 °C Density: 4.75 Storage Temperature: Ambient |
MDL Number:
MFCD00014166 CAS Number: 1303-36-2 EINECS: 215-119-5 UN: 3077 ADR: 9,III Merck Index: 14,805 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...