Barium sulfide ≥99.7% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Barium(II) sulfide, Barium(II) sulphide
12848-09
12848-18
CAAA12848-09EA
170.64
CAD
CAAA12848-09
CAAA12848-18
Barium sulfide ≥99.7% (metals basis)
Barium sulphide
MDL: MFCD00014192
Purity: 99.7%
CAS No.: 21109-95-5
Molecular Formula: BaS
Formula Weight: 169.40
Physical Form: -200 Mesh Powder
Density: 4.25
Appearance: Pale gray to yellow
Odor: Like rotten eggs
Melting Point: 1200°
Merck Reference: 14,995
UN No.: 3134
MDL No.: MFCD00014192
Formula:
BaS Melting Pt: 1200 °C Density: 4.25 Storage Temperature: Ambient |
MDL Number:
MFCD00014192 CAS Number: 21109-95-5 EINECS: 244-214-4 UN: 3134 ADR: 4.3,III Merck Index: 14,995 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...