Mercury selenide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

12832-03 12832-06
CAAA12832-03EA 127.82 CAD
CAAA12832-03 CAAA12832-06
Mercury selenide ≥99.999% (metals basis)
Mercury selenide

MDL: MFCD00049743


Purity: 99.999% CAS No.: 20601-83-6 Molecular Formula: HgSe Formula Weight: 279.55 Physical Form: -100 Mesh Powder Density: 8.21 Appearance: Gray Odor: Odorless Melting Point: subl. UN No.: 2025 MDL No.: MFCD00049743

Formula: HgSe
MW: 279.55
Melting Pt: 998 (subl.)
Density: 8.21
Storage Temperature: Ambient
MDL Number: MFCD00049743
CAS Number: 20601-83-6
EINECS: 243-910-5
UN: 2025
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR