Titanium (IV) sulfide ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

12826-06 12826-14
CAAA12826-06EA 515.08 CAD
CAAA12826-06 CAAA12826-14
Titanium (IV) sulfide ≥99.8% (metals basis)
Titanium (IV) sulphide

MDL: MFCD00049598


Purity: 99.8% CAS No.: 12039-13-3 Molecular Formula: TiS2 (approx.) Formula Weight: 112.03 Physical Form: -200 Mesh Powder Sensitivity: Moisture Sensitive UN No.: 3134 MDL No.: MFCD00049598

Formula: S₂Ti
Density: 3.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049598
CAS Number: 12039-13-3
EINECS: 234-883-0
UN: 3174
ADR: 4.2,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR