Boron ≥99.999% (metals basis), crystalline powder -4+40 mesh

Supplier: Thermo Scientific Chemicals
Warning

12820-03 12820-06
CAAA12820-03EA 474 CAD
CAAA12820-03 CAAA12820-06
Boron ≥99.999% (metals basis), crystalline powder -4+40 mesh
Boron

MDL: MFCD00134034


Purity: 99.999% CAS No.: 7440-42-8 Appearance: Brown to black Odor: Odorless UN No.: 3178 MDL No.: MFCD00134034

Formula: B
Storage Temperature: Ambient
MDL Number: MFCD00134034
CAS Number: 7440-42-8
EINECS: 231-151-2
UN: 3178
ADR: 4.1,III
Merck Index: 13,01333

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR