Copper ≥99.999% (metals basis), rods, Puratronic®, Ø 19 mm (0.8 in)
Supplier: Thermo Scientific Chemicals
12754-DJ
12754-DT
12754-HF
CAAA12754-DJEA
317.58
CAD
CAAA12754-DJ
CAAA12754-DT
CAAA12754-HF
Copper ≥99.999% (metals basis), rods, Puratronic®, Ø 19 mm (0.8 in)
Copper
MDL: MFCD00010965
Purity: 99.999%
CAS No.: 7440-50-8
Physical Form: ≈25.3g/cm
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...