Lead(II) iodide ≥99.9985% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead diiodide

12724-14 12724-22
CAAA12724-14EA 181.7 CAD
CAAA12724-14 CAAA12724-22
Lead(II) iodide ≥99.9985% (metals basis)
Lead (II) iodide

MDL: MFCD00011163


Purity: 99.9985% CAS No.: 10101-63-0 Molecular Formula: I2Pb Formula Weight: 461.00 Physical Form: Powder Density: 6.16 Appearance: Yellow Odor: Odorless Melting Point: 402° Boiling Point: 954° Merck Reference: 14,5411 UN No.: 2291 MDL No.: MFCD00011163

Formula: I₂Pb
MW: 461.01 g/mol
Boiling Pt: 954 °C
Melting Pt: 402 °C
Density: 6.16
Storage Temperature: Ambient
MDL Number: MFCD00011163
CAS Number: 10101-63-0
EINECS: 233-256-9
UN: 2291
ADR: 6.1,III
Merck Index: 14,5411

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR