Zinc ≥99.9998% (metals basis), rods, Ø 10 mm (0.39 in)
Supplier: Thermo Scientific Chemicals
12718-DJ
12718-DT
CAAA12718-DJEA
222.78
CAD
CAAA12718-DJ
CAAA12718-DT
Zinc ≥99.9998% (metals basis), rods, Ø 10 mm (0.39 in)
Zinc
MDL: MFCD00011291
Purity: 99.9998%
CAS No.: 7440-66-6
Physical Form: ≈5.60g/cm
Odor: Odorless
MDL No.: MFCD00011291
Formula:
Zn Boiling Pt: 908 °C (1013 hPa) Melting Pt: 420 °C Density: 7.14 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...