Silicon ≥99.5% (metals basis), crystalline powder -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

12681-09 12681-22 12681-36
CAAA12681-09EA 61.94 CAD
CAAA12681-09 CAAA12681-22 CAAA12681-36
Silicon ≥99.5% (metals basis), crystalline powder -325 mesh
Silicon

MDL: MFCD00085311


Purity: 99.5% CAS No.: 7440-21-3 Appearance: White Odor: Odorless UN No.: 1346 MDL No.: MFCD00085311

Formula: Si
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III
Merck Index: 12,08635

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR