Orthoboric acid ≥99.8%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
12680-A1
12680-A7
CAAA12680-A1EA
96.7
CAD
CAAA12680-A1
CAAA12680-A7
Orthoboric acid ≥99.8%
ortho-Boric acid
MDL: MFCD00011337
Beilstein Registry No.: 1697939
Fieser: 1,63 2,32 3,29 4,41 5,48 9,59 11,70
Purity: 99.8%
CAS No.: 10043-35-3
Molecular Formula: BH3O3
Formula Weight: 61.83
Flash Point: None
Physical Form: Granular
Density: 1.435
Appearance: White
Odor: Odorless
pH: 3.8-4.8
Melting Point: ca 185° dec.
Beilstein Reference: 1697939
Merck Reference: 14,1336
MDL No.: MFCD00011337
Formula:
B(OH)₃ Boiling Pt: 185 °C (1013 hPa) Melting Pt: ∼ 185 °C (dec.) Density: 1.435 Storage Temperature: Ambient |
MDL Number:
MFCD00011337 CAS Number: 10043-35-3 EINECS: 233-139-2 REACH: 01-2119486683-25 Merck Index: 14,1336 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...