Palladium (II) sulfide ≥99% (metals basis) Pd ≥ 72%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Palladium sulphide, Palladium monosulfide, Palladium monosulphide, Palladium sulfide
12674-03
12674-06
CAAA12674-03EA
398.16
CAD
CAAA12674-03
CAAA12674-06
Palladium (II) sulfide ≥99% (metals basis) Pd ≥ 72%
Palladium (II) sulphide
MDL: MFCD00016285
Purity: 99%
CAS No.: 12125-22-3
Molecular Formula: PdS
Formula Weight: 138.46
Physical Form: Crystalline
Density: 6.60
Melting Point: 950° dec.
MDL No.: MFCD00016285
Formula:
PdS Melting Pt: 950 °C (dec.) Density: 6,60 Storage Temperature: Ambient |
MDL Number:
MFCD00016285 CAS Number: 12125-22-3 EINECS: 235-190-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...