Diammonium hexabromoosmate (≥26,5% Os) ≥99.9% (metals basis) Os ≥ 26.5%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ammonium hexabromoosmate(IV)
12648-03
12648-06
CAAA12648-03EA
175.38
CAD
CAAA12648-03
CAAA12648-06
Diammonium hexabromoosmate (≥26,5% Os) ≥99.9% (metals basis) Os ≥ 26.5%
Diammonium hexabromoosmate
MDL: MFCD00049389
Purity: 99.9%
CAS No.: 24598-62-7
Molecular Formula: (NH4)2OsBr6
Formula Weight: 705.73
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Black
Odor: Odorless
MDL No.: MFCD00049389
Formula:
H₈Br₆N₂Os MW: 705.73 Storage Temperature: Ambient |
MDL Number:
MFCD00049389 CAS Number: 24598-62-7 EINECS: 246-340-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...