Potassium carbonate ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Warning

12609-14 12609-22 12609-36 12609-A1 12609-A7
CAAA12609-14EA 53.09 CAD
CAAA12609-14 CAAA12609-22 CAAA12609-36 CAAA12609-A1 CAAA12609-A7
Potassium carbonate ≥99.0% ACS
Potassium carbonate

MDL: MFCD00011382 Beilstein Registry No.: 4267587 Fieser: 1,1014 4,50 5,552 8,408 9,382 10,323 11,433 13,254


Purity: 99.0% min CAS No.: 584-08-7 Molecular Formula: K2CO3 Formula Weight: 138.21 Physical Form: Granular Density: 2.29 Melting Point: 891° Beilstein Reference: 4267587 Merck Reference: 14,7619 MDL No.: MFCD00011382

Formula: K₂CO₃
Melting Pt: 891 °C
Density: 2.29
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 14,7619
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matter 0.01%
Cl 0.3%
PO6 0.001%
SiO2 0.005%
SO3 0.004%
Ca 0.005%
MG 0.002%
Heavy metals (as Pb) 5 ppm
Fe 5 ppm
NA 0.0002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR