Tellurium ≥99.9999% (metals basis), broken ingot, Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
12607-14
12607-22
12607-36
CAAA12607-14EA
320.74
CAD
CAAA12607-14
CAAA12607-22
CAAA12607-36
Tellurium ≥99.9999% (metals basis), broken ingot, Puratronic®
Tellurium
MDL: MFCD00134062
Purity: 99.9999+%
CAS No.: 13494-80-9
Odor: Odorless
MDL No.: MFCD00134062
Formula:
Te Boiling Pt: 990 °C (1013 hPa) Melting Pt: 450 °C Density: 8.83 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134062 CAS Number: 13494-80-9 EINECS: 236-813-4 UN: 3284 ADR: 6.1,III Merck Index: 12,09273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...