Lead ≥99.8% (metals basis), foil, Thickness: 1.19 mm (0.0469 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12604-RF
12604-VA
12604-VM
CAAA12604-RFEA
140.46
CAD
CAAA12604-RF
CAAA12604-VA
CAAA12604-VM
Lead ≥99.8% (metals basis), foil, Thickness: 1.19 mm (0.0469 in)
Lead
MDL: MFCD00134050
Notes: Due to the nature of this material, there may be fluctuations in the thickness measurements of this foil.
Purity: 99.8%
CAS No.: 7439-92-1
Physical Form: ≈277g/15x15cm
Appearance: Grey
Odor: Odorless
MDL No.: MFCD00134050
Formula:
Pb Boiling Pt: 1740 °C (1013 hPa) Melting Pt: 327 °C Density: 11.3437 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134050 CAS Number: 7439-92-1 EINECS: 231-100-4 Merck Index: 12,05410 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...