Lithium metaborate, anhydrous ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
12591-18
12591-30
12591-A1
CAAA12591-18EA
98.43
CAD
CAAA12591-18
CAAA12591-30
CAAA12591-A1
Lithium metaborate, anhydrous ≥99.9% (metals basis)
Lithium metaborate
MDL: MFCD00011089
Purity: 99.9%
CAS No.: 13453-69-5
Molecular Formula: BLiO2
Formula Weight: 49.75
Physical Form: Powder
Density: 1.397
Appearance: White
Odor: Odorless
Melting Point: 845°
MDL No.: MFCD00011089
Formula:
BLiO₂ Boiling Pt: 1719 °C (1013 hPa) Melting Pt: 845 °C Density: 1.397 Storage Temperature: Ambient |
MDL Number:
MFCD00011089 CAS Number: 13453-69-5 EINECS: 236-631-5 Merck Index: 13,05546 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...