Sodium selenite, anhydrous ≥99% (metals basis) typ. ≥ 99.75%
Supplier: Thermo Scientific Chemicals
|
Danger
|
12585-22
12585-36
CAAA12585-22EA
142.2
CAD
CAAA12585-22
CAAA12585-36
Sodium selenite, anhydrous ≥99% (metals basis) typ. ≥ 99.75%
Sodium selenite
In glass manufacturing
Purity: 99% min
CAS No.: 10102-18-8
Molecular Formula: Na2O3Se
Formula Weight: 172.94
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
pH: 9
Melting Point:
In glass manufacturing
Formula:
Na₂SeO₃ Melting Pt: >350 °C Density: 3.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003489 CAS Number: 10102-18-8 EINECS: 233-267-9 UN: 2630 ADR: 6.1,I Merck Index: 14,8673 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...