Palladium ≥99.95% (metals basis), rods, Ø 6.35 mm

Supplier: Thermo Scientific Chemicals

12557-DH 12557-DM
CAAA12557-DHEA 4332.36 CAD
CAAA12557-DH CAAA12557-DM
Palladium ≥99.95% (metals basis), rods, Ø 6.35 mm
Palladium

MDL: MFCD00011167


Purity: 99.95% CAS No.: 7440-05-3 Physical Form: ≈3.80g/cm Odor: Odorless MDL No.: MFCD00011167

Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR