Aluminium acetylacetonate
Supplier: Thermo Scientific Chemicals
|
Danger
|
12542-22
12542-36
CAAA12542-22EA
61.62
CAD
CAAA12542-22
CAAA12542-36
Aluminium acetylacetonate
Aluminium acetylacetonate
MDL: MFCD00000013
Beilstein Registry No.: 4157942
CAS No.: 13963-57-0
Molecular Formula: C15H21AlO6
Formula Weight: 324.31
Physical Form: Powder
Density: 1.27
Melting Point: 192-194°
Boiling Point: 314-316°
Beilstein Reference: 4157942
UN No.: 3467
MDL No.: MFCD00000013
Formula:
C₁₅H₂₁AlO₆ Boiling Pt: 314…316 °C Melting Pt: 192…194 °C Density: 1,27 Storage Temperature: Ambient |
MDL Number:
MFCD00000013 CAS Number: 13963-57-0 EINECS: 237-741-6 UN: 3467 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...