Chromium(III) acetylacetonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12538-22
12538-36
CAAA12538-22EA
89.43
CAD
CAAA12538-22
CAAA12538-36
Chromium(III) acetylacetonate 97%
Chromium (III) acetylacetonate
Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVD
Purity: 97%
CAS No.: 21679-31-2
Molecular Formula: C15H21CrO6
Formula Weight: 349.33
Physical Form: Crystalline
Density: 1.35
Melting Point: 216°
Boiling Point: 340°
Solubility: Insoluble in water. Soluble in toluene, acetic acid
MDL No.: MFCD00000015
Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVD
Formula:
C₁₅H₂₁CrO₆ Boiling Pt: 340 °C Melting Pt: 216 °C Density: 1,35 Storage Temperature: Ambient |
MDL Number:
MFCD00000015 CAS Number: 21679-31-2 EINECS: 244-526-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...