Cobalt(II) acetylacetonate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobalt(II) bis(acetylacetonate), Cobalt (II) bis(2,4-pentanedionato-O,O'), Cobalt (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')cobalt (II)

12537-18 12537-30
CAAA12537-18EA 102.07 CAD
CAAA12537-18 CAAA12537-30
Cobalt(II) acetylacetonate
Cobalt (II) acetylacetonate

MDL: MFCD00000014


CAS No.: 14024-48-7 Molecular Formula: C10H14CoO4 Formula Weight: 257.14 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Blue/purple Odor: Odorless Melting Point: 165-170° MDL No.: MFCD00000014

Formula: C₁₀H₁₄CoO₄
MW: 257.15 g/mol
Melting Pt: 165…170 °C
Density: 0.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000014
CAS Number: 14024-48-7
EINECS: 237-855-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR