Cobalt (III) acetylacetonate
Supplier: Thermo Scientific Chemicals
|
Warning
|
12536-06
12536-14
12536-22
CAAA12536-06EA
49.45
CAD
CAAA12536-06
CAAA12536-14
CAAA12536-22
Cobalt (III) acetylacetonate
Cobalt (III) acetylacetonate
MDL: MFCD00013488
CAS No.: 21679-46-9
Molecular Formula: C15H21CoO6
Formula Weight: 356.26
Physical Form: Powder
Sensitivity: Moisture Sensitive and Hygroscopic
Odor: Light
MDL No.: MFCD00013488
Formula:
C₁₅H₂₁CoO₆ Melting Pt: 211 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013488 CAS Number: 21679-46-9 EINECS: 244-527-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...