Copper(II) acetylacetonate, hydrate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate

12535-14 12535-22 12535-36
CAAA12535-14EA 49.45 CAD
CAAA12535-14 CAAA12535-22 CAAA12535-36
Copper(II) acetylacetonate, hydrate
Copper (II) acetylacetonate

MDL: MFCD00000016


CAS No.: 13395-16-9 Molecular Formula: C10H14CuO4.xH2O Formula Weight: 261.76anhy Physical Form: Powder Odor: Weak Melting Point: 230° MDL No.: MFCD00000016

Formula: C₁₀H₁₄CuO₄
MW: 261.76 g/mol
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00000016
CAS Number: 13395-16-9
EINECS: 236-477-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR