Iron(III) acetylacetonate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')

12534-22 12534-36
CAAA12534-22EA 101.59 CAD
CAAA12534-22 CAAA12534-36
Iron(III) acetylacetonate
Iron (III) acetylacetonate

MDL: MFCD00000020 Beilstein Registry No.: 4157960 Fieser: 4,268 6,304 18,196


CAS No.: 14024-18-1 Molecular Formula: C15H21FeO6 Formula Weight: 353.17 Physical Form: Powder Appearance: Dark red Melting Point: ca 183° dec. Beilstein Reference: 4157960 MDL No.: MFCD00000020

Formula: C₁₅H₂₁FeO₆
MW: 353.18 g/mol
Boiling Pt: 100 °C (9,7513 mmHg)
Melting Pt: ∼ 183 °C (dec.)
Density: 5,24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000020
CAS Number: 14024-18-1
EINECS: 237-853-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR