Manganese(III) acetylacetonate
Supplier: Thermo Scientific Chemicals
12530-09
12530-18
12530-30
CAAA12530-09EA
53.88
CAD
CAAA12530-09
CAAA12530-18
CAAA12530-30
Manganese(III) acetylacetonate
Manganese (III) acetylacetonate
MDL: MFCD00000023
Soluble in benzene, ethyl acetate
CAS No.: 14284-89-0
Molecular Formula: C15H21MnO6
Formula Weight: 352.24
Physical Form: Powder
Sensitivity: Hygroscopic
Melting Point: ≈160° dec.
MDL No.: MFCD00000023
Formula:
C₁₅H₂₁MnO₆ Melting Pt: ∼160 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00000023 CAS Number: 14284-89-0 EINECS: 238-188-3 |
Specification Test Results
Form | Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...