Cobalt(II) hydroxide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cobalt dihydroxide, Cobaltous hydroxide, Cobalt(II) hydroxide
MDL: MFCD00016015
Soluble in acids, ammonia. Very slightly soluble in water
Purity: 99.9%
CAS No.: 21041-93-0
Molecular Formula: H2CoO2
Formula Weight: 92.95
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: Pink to purple
Odor: Odorless
Melting Point: 250° dec.
Solubility: Soluble in acids, ammonia. Very slightly soluble in water
Merck Reference: 14,2442
MDL No.: MFCD00016015
Formula:
H₂CoO₂ MW: 92.95 g/mol Melting Pt: 250 °C (dec.) Density: 3.597 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016015 CAS Number: 21041-93-0 EINECS: 244-166-4 Merck Index: 14,2442 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...