Chromium(VI) oxide 99%

Supplier: Thermo Scientific Chemicals
Danger

12522-30 12522-A1 12522-D9
CAAA12522-30EA 64.94 CAD
CAAA12522-30 CAAA12522-A1 CAAA12522-D9
Chromium(VI) oxide 99%
Chromium (VI) oxide

MDL: MFCD00010952


Purity: 99% CAS No.: 1333-82-0 Molecular Formula: CrO3 Formula Weight: 99.99 Physical Form: Crystalline Density: 2.7 Appearance: red Melting Point: 196° Boiling Point: 250° dec. Merck Reference: 14,2235 UN No.: 1463 MDL No.: MFCD00010952

Formula: CrO₃
Boiling Pt: 250 °C (dec.)
Melting Pt: 196 °C
Density: 2.7
Storage Temperature: Ambient
MDL Number: MFCD00010952
CAS Number: 1333-82-0
EINECS: 215-607-8
UN: 1463
ADR: 5.1,II
REACH: 01-2119458868-17
Merck Index: 14,2235

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR