Nickel (II) sulfate hexahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

12514-36 12514-A3
CAAA12514-36EA 113.6 CAD
CAAA12514-36 CAAA12514-A3
Nickel (II) sulfate hexahydrate 98%
Nickel sulphate hexahydrate

In nickel plating, as mordant in dyeing and in printing textiles


Purity: 98% CAS No.: 10101-97-0 Molecular Formula: NiO4S.6H2O Formula Weight: 262.86 (154.77anhy) Physical Form: Crystalline Density: 2.070 Appearance: Green Odor: Odorless pH: 4.3-4.7 Solubility: Soluble in water. Slightly soluble in alcohol, methanol Merck Reference: 14,6517 UN No.: 3288 MDL No.: MFCD00149813


In nickel plating, as mordant in dyeing and in printing textiles

Formula: NiSO₄·6H₂O
Melting Pt: 840 °C
Density: 2.070
Storage Temperature: Ambient
MDL Number: MFCD00149813
CAS Number: 10101-97-0
EINECS: 232-104-9
UN: 3288
ADR: 6.1,III
Merck Index: 14,6517

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR