Strontium oxide, tech.

Supplier: Thermo Scientific Chemicals
Danger

12495-30 12495-A1
CAAA12495-30EA 176.96 CAD
CAAA12495-30 CAAA12495-A1 CAAA12495-A7
Strontium oxide, tech.
Strontium oxide

In the manufacture of strontium salts, pyrotechnics, and pigments, in greases and soaps, and as a desiccant


CAS No.: 1314-11-0 Molecular Formula: SrO, may also contain Sr(OH)2, SrCO3 Formula Weight: 103.62 Physical Form: -100 Mesh Powder Density: 4.700 Sensitivity: Moisture Sensitive Boiling Point: ca 3000° Solubility: Soluble in fused potassium hydroxide. Insoluble in ether and acetone. Decomposes in water to Sr(OH)2 with evolution of heat Merck Reference: 14,8846 UN No.: 3262 MDL No.: MFCD00049558


In the manufacture of strontium salts, pyrotechnics, and pigments, in greases and soaps, and as a desiccant

Formula: SrO
Boiling Pt: ∼3000 °C
Melting Pt: 2430 °C
Density: 4.700
Storage Temperature: Ambient
MDL Number: MFCD00049558
CAS Number: 1314-11-0
EINECS: 215-219-9
UN: 3262
ADR: 8,II
Merck Index: 14,8846

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR